Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cyp27c1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113337.3 | NP_001106808.2 | 8 - 9 | 1636 | 77 | |
GenBank mRNA | BC154632.1 | - | 8 - 9 | 1545 | 77 | |
BC171576.1 | - | 8 - 9 | 1321 | 77 | ||
BC171578.1 | - | 8 - 9 | 1321 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |