Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pi4kab | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113342.1 | NP_001106813.1 | 5 - 6 | 526 | 60 | |
GenBank mRNA | BC155592.1 | - | 5 - 6 | 526 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |