Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: keap1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113477.1 | NP_001106948.1 | 6 - 7 | 1882 | 63 | |
XM_005168685.3 | XP_005168742.1 | 6 - 7 | 2030 | 63 | ||
XM_021476739.1 | XP_021332414.1 | 6 - 7 | 1920 | 63 | ||
GenBank mRNA | AB271119.1 | - | 6 - 7 | 1882 | 63 | |
BC155079.1 | - | 6 - 7 | 1906 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |