Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: kiss1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113489.1 | NP_001106961.1 | 1 - 2 | 36 | 103 | |
GenBank mRNA | AB245404.1 | - | 1 - 2 | 36 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Danio rerio Probe spans exons |