Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: anapc7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113585.1 | NP_001107057.1 | 3 - 4 | 414 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Danio rerio Probe spans exons |