Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tfb2m | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113617.2 | NP_001107089.2 | 7 - 8 | 1105 | 104 | |
GenBank mRNA | BC154809.1 | - | 7 - 8 | 104 | ||
BC171547.1 | - | 7 - 8 | 1028 | 104 | ||
EH461916.1 | - | 2 - 3 | 832 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |