Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sgk2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113622.1 | NP_001107094.1 | 6 - 7 | 530 | 130 | |
XM_009296592.3 | XP_009294867.1 | 6 - 7 | 590 | 130 | ||
GenBank mRNA | BC154535.1 | - | 6 - 7 | 530 | 130 | |
BC171486.1 | - | 6 - 7 | 497 | 130 | ||
BC171490.1 | - | 6 - 7 | 497 | 130 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |