Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lifrb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113732.1 | NP_001107204.1 | 13 - 14 | 1922 | 112 | |
XM_009295324.3 | XP_009293599.1 | 13 - 14 | 1992 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Danio rerio Probe spans exons |