Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: il6r | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001330258.2 | NP_001317187.2 | 4 - 5 | 1447 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Danio rerio Probe spans exons |