Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: slc4a2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114440.1 | NP_001107912.1 | 15 - 16 | 2506 | 84 | |
GenBank mRNA | DQ286970.1 | - | 15 - 16 | 2506 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Danio rerio Probe spans exons |