Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hoxd13a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131169.3 | NP_571244.2 | 1 - 2 | 609 | 64 | |
GenBank mRNA | BC091996.1 | - | 1 - 2 | 613 | 64 | |
BC153652.1 | - | 1 - 2 | 622 | 64 | ||
BC163878.1 | - | 1 - 2 | 593 | 64 | ||
BC165451.1 | - | 1 - 2 | 572 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |