Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tbx18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153665.1 | NP_705951.1 | 6 - 7 | 1122 | 87 | |
GenBank mRNA | AF448503.3 | - | 6 - 7 | 1110 | 87 | |
BC154826.1 | - | 6 - 7 | 1106 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |