Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tbx6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153666.2 | NP_705952.2 | 7 - 8 | 1206 | 110 | |
GenBank mRNA | AB036516.1 | - | 7 - 8 | 1206 | 110 | |
BC163297.1 | - | 7 - 8 | 1094 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |