Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: selenow2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_194418.3 | NP_919399.3 | 2 - 3 | 251 | 81 | |
GenBank mRNA | AY216583.1 | - | 2 - 3 | 152 | 81 | |
CK687346.1 | - | 2 - 3 | 221 | 81 | ||
EH115335.1 | - | 2 - 3 | 233 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Danio rerio Probe spans exons |