Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: oaz1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_194432.3 | NP_919413.2 | 3 - 4 | 523 | 64 | |
GenBank mRNA | AB017118.1 | - | 3 - 4 | 480 | 64 | |
BC056833.1 | - | 3 - 4 | 496 | 64 | ||
CF550492.1 | - | 3 - 4 | 558 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |