Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: p2rx4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198987.2 | NP_945338.2 | 10 - 11 | 1069 | 89 | |
XM_021478206.1 | XP_021333881.1 | 10 - 11 | 1521 | 89 | ||
GenBank mRNA | AY292652.1 | - | 10 - 11 | 1064 | 89 | |
BC162774.1 | - | 10 - 11 | 1069 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |