Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cyp4t8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199216.2 | NP_954686.2 | 6 - 7 | 828 | 73 | |
GenBank mRNA | BC056734.1 | - | 6 - 7 | 796 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |