Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lnpa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199480.1 | NP_955774.1 | 7 - 8 | 639 | 90 | |
GenBank mRNA | AY423028.1 | - | 7 - 8 | 568 | 90 | |
BC050510.1 | - | 7 - 8 | 639 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |