Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199598.1 | NP_955892.1 | 2 - 3 | 206 | 70 | |
GenBank mRNA | BC045362.1 | - | 2 - 3 | 206 | 70 | |
BC165743.1 | - | 2 - 3 | 136 | 70 | ||
BC166040.1 | - | 2 - 3 | 202 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |