Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mob1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199914.1 | NP_956208.1 | 3 - 4 | 571 | 116 | |
XM_021475960.1 | XP_021331635.1 | 3 - 4 | 659 | 116 | ||
GenBank mRNA | BC045979.1 | - | 3 - 4 | 571 | 116 | |
BC066567.1 | - | 3 - 4 | 569 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |