Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: stx4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200221.1 | NP_956515.1 | 3 - 4 | 389 | 83 | |
GenBank mRNA | BC046039.1 | - | 3 - 4 | 389 | 83 | |
BC152678.1 | - | 3 - 4 | 469 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |