Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nol12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200290.1 | NP_956584.1 | 4 - 5 | 440 | 69 | |
GenBank mRNA | BC049449.1 | - | 4 - 5 | 440 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |