Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pex13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200645.1 | NP_956939.1 | 2 - 3 | 931 | 125 | |
GenBank mRNA | BC044529.1 | - | 2 - 3 | 1975 | 125 | |
BC057464.1 | - | 2 - 3 | 931 | 125 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |