Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: grap2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200678.2 | NP_956972.1 | 5 - 6 | 614 | 74 | |
GenBank mRNA | BC058307.1 | - | 520 | 74 | ||
BC066707.1 | - | 5 - 6 | 494 | 74 | ||
EH590682.1 | - | 5 - 6 | 635 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |