Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pam16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200804.2 | NP_957098.2 | 4 - 5 | 442 | 68 | |
GenBank mRNA | BC059670.1 | - | 4 - 5 | 390 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |