Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ccl44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001115102.1 | NP_001108574.1 | 4 - 5 | 387 | 62 | |
XM_009305799.3 | XP_009304074.1 | 4 - 5 | 465 | 62 | ||
GenBank mRNA | AB331768.1 | - | 4 - 5 | 387 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Danio rerio Probe spans exons |