Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dpp6b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001115122.1 | NP_001108594.1 | 29 - 30 | 2869 | 82 | |
XM_005163289.3 | XP_005163346.1 | 24 - 25 | 2837 | 82 | ||
XM_005163290.4 | XP_005163347.1 | 24 - 25 | 2312 | 82 | ||
XM_005163291.4 | XP_005163348.1 | 24 - 25 | 2280 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Danio rerio Probe spans exons |