Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: prom1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001115143.2 | NP_001108615.2 | 13 - 14 | 1756 | 71 | |
XM_005157347.3 | XP_005157404.1 | 13 - 14 | 1881 | 71 | ||
XM_005157349.3 | XP_005157406.1 | 15 - 16 | 1933 | 71 | ||
XM_005157351.3 | XP_005157408.1 | 14 - 15 | 1909 | 71 | ||
XM_009291267.3 | XP_009289542.1 | 15 - 16 | 2035 | 71 | ||
XM_009291269.3 | XP_009289544.1 | 13 - 14 | 1881 | 71 | ||
XM_021480968.1 | XP_021336643.1 | 15 - 16 | 2036 | 71 | ||
XM_021480969.1 | XP_021336644.1 | 14 - 15 | 2012 | 71 | ||
XM_021480970.1 | XP_021336645.1 | 14 - 15 | 2009 | 71 | ||
XM_021480971.1 | XP_021336646.1 | 15 - 16 | 2035 | 71 | ||
XM_021480972.1 | XP_021336647.1 | 15 - 16 | 2035 | 71 | ||
XM_021480973.1 | XP_021336648.1 | 13 - 14 | 1882 | 71 | ||
XM_021480974.1 | XP_021336649.1 | 15 - 16 | 2036 | 71 | ||
XM_021480975.1 | XP_021336650.1 | 15 - 16 | 2035 | 71 | ||
GenBank mRNA | AF160970.1 | - | 13 - 14 | 1881 | 71 | |
BC163446.1 | - | 13 - 14 | 1756 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |