Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: prom1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001115143.2 | NP_001108615.2 | 6 - 7 | 861 | 65 | |
XM_005157347.3 | XP_005157404.1 | 6 - 7 | 986 | 65 | ||
XM_005157351.3 | XP_005157408.1 | 7 - 8 | 1014 | 65 | ||
XM_009291269.3 | XP_009289544.1 | 6 - 7 | 986 | 65 | ||
XM_021480969.1 | XP_021336644.1 | 7 - 8 | 1117 | 65 | ||
XM_021480973.1 | XP_021336648.1 | 6 - 7 | 987 | 65 | ||
GenBank mRNA | AF160970.1 | - | 6 - 7 | 986 | 65 | |
BC163446.1 | - | 6 - 7 | 861 | 65 | ||
HQ386793.1 | - | 7 - 8 | 1018 | 65 | ||
HQ386795.1 | - | 6 - 7 | 986 | 65 | ||
HQ386796.1 | - | 6 - 7 | 998 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Probe spans exons |