Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: prom1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001115143.2 | NP_001108615.2 | 8 - 9 | 1154 | 69 | |
XM_005157347.3 | XP_005157404.1 | 8 - 9 | 1279 | 69 | ||
XM_005157349.3 | XP_005157406.1 | 10 - 11 | 1331 | 69 | ||
XM_005157351.3 | XP_005157408.1 | 9 - 10 | 1307 | 69 | ||
XM_009291267.3 | XP_009289542.1 | 10 - 11 | 1433 | 69 | ||
XM_009291269.3 | XP_009289544.1 | 8 - 9 | 1279 | 69 | ||
XM_021480968.1 | XP_021336643.1 | 10 - 11 | 1434 | 69 | ||
XM_021480969.1 | XP_021336644.1 | 9 - 10 | 1410 | 69 | ||
XM_021480970.1 | XP_021336645.1 | 9 - 10 | 1407 | 69 | ||
XM_021480971.1 | XP_021336646.1 | 10 - 11 | 1433 | 69 | ||
XM_021480972.1 | XP_021336647.1 | 10 - 11 | 1433 | 69 | ||
XM_021480973.1 | XP_021336648.1 | 8 - 9 | 1280 | 69 | ||
XM_021480974.1 | XP_021336649.1 | 10 - 11 | 1434 | 69 | ||
XM_021480975.1 | XP_021336650.1 | 10 - 11 | 1433 | 69 | ||
GenBank mRNA | AF160970.1 | - | 8 - 9 | 1279 | 69 | |
BC163446.1 | - | 8 - 9 | 1154 | 69 | ||
HQ386796.1 | - | 8 - 9 | 1291 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Probe spans exons |