Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tmtopsa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001118899.2 | NP_001112371.1 | 1 - 2 | 432 | 60 | |
GenBank mRNA | BC163681.1 | - | 1 - 2 | 432 | 60 | |
KT008411.1 | - | 1 - 2 | 368 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 G-protein coupled receptor Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |