Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: slc6a22.2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001122761.1 | NP_001116233.1 | 2 - 3 | 393 | 68 | |
XM_005166045.4 | XP_005166102.1 | 2 - 3 | 406 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Danio rerio Probe spans exons |