Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: slc6a22.2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001122761.1 | NP_001116233.1 | 4 - 5 | 673 | 98 | |
XM_005166045.4 | XP_005166102.1 | 4 - 5 | 686 | 98 | ||
XM_005166046.4 | XP_005166103.1 | 5 - 6 | 595 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Danio rerio Probe spans exons |