Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lhx4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001122973.1 | NP_001116445.1 | 4 - 5 | 611 | 57 | |
XM_021478301.1 | XP_021333976.1 | 4 - 5 | 645 | 57 | ||
XM_021478302.1 | XP_021333977.1 | 4 - 5 | 637 | 57 | ||
GenBank mRNA | BC129374.1 | - | 4 - 5 | 780 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |