Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: slc37a4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001328095.1 | NP_001315024.1 | 2 - 3 | 916 | 110 | |
XM_005169205.4 | XP_005169262.1 | 2 - 3 | 462 | 110 | ||
XM_009305596.3 | XP_009303871.1 | 2 - 3 | 494 | 110 | ||
XM_021479139.1 | XP_021334814.1 | 2 - 3 | 586 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Danio rerio Probe spans exons |