Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nek2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201050.1 | NP_957344.1 | 8 - 9 | 1180 | 67 | |
GenBank mRNA | BC048055.1 | - | 8 - 9 | 1180 | 67 | |
BC164531.1 | - | 7 - 8 | 1078 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |