Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dtnbp1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201134.1 | NP_957428.1 | 3 - 4 | 223 | 84 | |
XM_005158097.3 | XP_005158154.1 | 3 - 4 | 490 | 84 | ||
XM_005158098.4 | XP_005158155.1 | 3 - 4 | 488 | 84 | ||
XM_005158099.4 | XP_005158156.1 | 3 - 4 | 487 | 84 | ||
XM_005158100.3 | XP_005158157.1 | 3 - 4 | 491 | 84 | ||
XM_005158101.3 | XP_005158158.1 | 3 - 4 | 490 | 84 | ||
XM_021466652.1 | XP_021322327.1 | 3 - 4 | 493 | 84 | ||
GenBank mRNA | BC049451.1 | - | 3 - 4 | 223 | 84 | |
BC152672.1 | - | 3 - 4 | 461 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |