Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ndel1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201307.2 | NP_958464.1 | 7 - 8 | 880 | 74 | |
GenBank mRNA | BC044386.2 | - | 879 | 74 | ||
BC165239.1 | - | 6 - 7 | 827 | 74 | ||
EF133515.1 | - | 7 - 8 | 823 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |