Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tmsb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205581.1 | NP_991144.1 | 2 - 3 | 169 | 130 | |
GenBank mRNA | AF006831.1 | - | 2 - 3 | 169 | 130 | |
BC098515.1 | - | 2 - 3 | 194 | 130 | ||
BC115156.1 | - | 2 - 3 | 193 | 130 | ||
BC159220.1 | - | 166 | 130 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |