Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: grnb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212738.1 | NP_997903.1 | 11 - 12 | 1257 | 87 | |
XM_021470114.1 | XP_021325789.1 | 11 - 12 | 1580 | 87 | ||
GenBank mRNA | AY289606.1 | - | 11 - 12 | 1257 | 87 | |
BC096854.1 | - | 11 - 12 | 1251 | 87 | ||
BC163965.1 | - | 9 - 10 | 1164 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |