Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: chmp2bb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212904.2 | NP_998069.1 | 4 - 5 | 565 | 75 | |
GenBank mRNA | BC067142.1 | - | 4 - 5 | 519 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |