Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: chmp2bb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212904.2 | NP_998069.1 | 5 - 6 | 662 | 68 | |
GenBank mRNA | BC067142.1 | - | 5 - 6 | 616 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |