Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: itm2bb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212976.1 | NP_998141.1 | 2 - 3 | 278 | 78 | |
GenBank mRNA | BC053278.1 | - | 2 - 3 | 278 | 78 | |
BC155109.1 | - | 2 - 3 | 275 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |