Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rps11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213377.1 | NP_998542.1 | 2 - 3 | 175 | 125 | |
GenBank mRNA | AY648802.1 | - | 2 - 3 | 173 | 125 | |
BC046054.1 | - | 2 - 3 | 175 | 125 | ||
BC159194.1 | - | 2 - 3 | 175 | 125 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Danio rerio Probe spans exons |