Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hnrnpl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213383.1 | NP_998548.1 | 6 - 7 | 743 | 65 | |
XM_005169693.4 | XP_005169750.1 | 6 - 7 | 764 | 65 | ||
GenBank mRNA | BC054655.1 | - | 6 - 7 | 743 | 65 | |
BC165353.1 | - | 6 - 7 | 731 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Probe spans exons |