Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: vcanb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214688.1 | NP_999853.1 | 13 - 14 | 4446 | 78 | |
GenBank mRNA | AB119257.1 | - | 13 - 14 | 4446 | 78 | |
BC162605.1 | - | 13 - 14 | 4432 | 78 |
Gene Symbol: LOC103909969 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_009298338.3 | XP_009296613.1 | 1 - 2 | 210 | 78 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |