Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OASL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081797.1 | NP_001075266.1 | 4 - 5 | 910 | 93 | |
XM_005612545.3 | XP_005612602.1 | 1544 | 93 | |||
GenBank mRNA | AY463162.1 | - | 4 - 5 | 910 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Equus caballus Probe spans exons |