Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL1R2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081816.1 | NP_001075285.1 | 6 - 7 | 875 | 105 | |
XM_005599763.3 | XP_005599820.2 | 1041 | 105 | |||
XM_005599765.3 | XP_005599822.1 | 937 | 105 | |||
XM_023618377.1 | XP_023474145.1 | 922 | 105 | |||
GenBank mRNA | AB033415.1 | - | 6 - 7 | 875 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Equus caballus Probe spans exons |