Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MMP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081847.2 | NP_001075316.1 | 4 - 5 | 679 | 70 | |
GenBank mRNA | AF148882.1 | - | 4 - 5 | 681 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Equus caballus Inventoried Probe spans exons |