Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRISP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081874.1 | NP_001075343.1 | 4 - 5 | 330 | 104 | |
XM_014734198.2 | XP_014589684.2 | 382 | 104 | |||
GenBank mRNA | AJ001400.1 | - | 4 - 5 | 330 | 104 | |
AJ459964.1 | - | 4 - 5 | 330 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Equus caballus Probe spans exons |